Learn More
HOMER2, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human HOMER2 protein.
Brand: Abnova H00009455-D01.100uL
Description
This gene encodes a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function. The encoded protein may be involved in cell growth. Four transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: MGEQPIFTTRAHVFQIDPNTKKNWMPASKQAVTVSYFYDVTRNSYRIISVDGAKVIINSTITPNMTFTKTSQKFGQWADSRANTVFGLGFSSEQQLTKFAEKFQEVKEAAKIAKDKTQEKIETSSNHSQASSVNGTDDEKASHAGPANTHLKSENDKLKIALTQSAANVKKWEIELQTLRESNARLTTALQESAASVEQWKRQFSICRDENDRLRNKIDELEEQCSEINREKEKNTQLKRRIEELEAELREKETELKDLRKQSEIIPQLMSECEYVSEKLEAAERDNQNLEDKVRSLKTDIEESKYRQRHLKVELKSFLEVLDGKIDDLHDFRRGLSKLGTDNSpecifications
| HOMER2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human HOMER2 protein. | |
| HOMER2 | |
| ACPD/CPD/HOMER-2/HOMER2A/HOMER2B/Vesl-2 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| No additive | |
| NM_004839.2 | |
| HOMER2 | |
| HOMER2 (NP_004830.2, 1 a.a. ∼ 343 a.a) full-length human protein. | |
| RUO | |
| 9455 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.