missing translation for 'onlineSavingsMsg'
Learn More

Anti-Human CD14 (26IC), Proteintech

Product Code. p-200108229 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235292 100 μg 100µg
30233926 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235292 Supplier Proteintech Supplier No. 652461IG100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CD14 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD14. This antibody recognizes human antigen. The CD14 antibody has been validated for the following applications: FC, ELISA analysis.

Specifications

Antigen CD14
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 26IC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 929
Target Species Human
Content And Storage Store at 2-8°C. Stable for one year after shipment.
Product Type Flow Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.