missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD20 (2H7), Proteintech
Mouse Monoclonal Antibody
€ 40.00 - € 125.00
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 2H7 |
| Concentration | 500 μg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236602
|
Proteintech
65085-1-IG-500UG |
500 μg |
€ 125.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235617
|
Proteintech
65085-1-IG-100UG |
100 μg |
€ 40.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD20 antibody from Proteintech is a mouse monoclonal antibody generated with human tonsillar b cells of human CD20. This antibody recognizes human antigen. The CD20 antibody has been validated for the following applications: FC analysis.Specifications
| CD20 | |
| 500 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 931 | |
| Human tonsillar B cells | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
| 2H7 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| B1; Bp35; CD20; LEU-16; MGC3969; MS4A2; S7 | |
| IgG2b κ | |
| Affinity purification | |
| Flow Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title