Learn More
Anti-Human CD226 (11A8), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65247-1-IG-100UG
 View more versions of this product
Product Code. 30234695
  • € 64.00 / 100µg
Description
The CD226 antibody from Proteintech is a mouse monoclonal antibody generated with human nk cells of human CD226. This antibody recognizes human antigen. The CD226 antibody has been validated for the following applications: FC, ELISA analysis.
Specifications
Specifications
CD226
Monoclonal
0.5 mg/mL
DNAM-1; DNAM1; PTA1; TLiSA1
Human NK Cells
100 μg
Primary
Human
Flow Antibody
IgG1 κ
Flow Cytometry, ELISA
11A8
Unconjugated
Mouse
Affinity purification
RUO
10666
Store at 2-8°C. Stable for one year after shipment.