missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD28 (CD28.2), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65099-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD28 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD28. This antibody recognizes human antigen. The CD28 antibody has been validated for the following applications: FC analysis.Specifications
| CD28 | |
| Monoclonal | |
| 0.5 mg/mL | |
| MGC138290; Tp44 | |
| Affinity purification | |
| RUO | |
| 940 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| CD28.2 | |
| Unconjugated | |
| Mouse | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction