Learn More
CD4 Mouse anti-Human, Brilliant Violet 605, Clone: RPA-T4, BD
Mouse Monoclonal Antibody
€ 215.73 - € 489.80
Specifications
| Antigen | CD4 |
|---|---|
| Clone | RPA-T4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 605 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15862119
|
BD Biosciences
562659 |
25 Tests |
€ 216.00 € 215.73 / Each Save € 0.27 0.13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15852119
|
BD Biosciences
562658 |
100 Tests |
€ 505.00 € 489.80 / Each Save € 15.20 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RPA-T4 monoclonal antibody specifically binds to CD4, a 59 kDa single-chain transmembrane glycoprotein that is expressed on T-helper/inducer cell populations. CD4 is also expressed on thymocytes subsets and at lower levels on monocytes and macrophages. CD4 functions as a co-receptor in MHC class II-restricted antigen-induced T cell activation and as a receptor for human immunodeficiency viruses (HIV). This antibody binds to the D1 domain (CDR1 and CDR3 epitopes) of the CD4 antigen and reacts with approximately 80% of thymocytes and 45% of peripheral blood lymphocytes. RPA-T4 is capable of blocking HIV-1, gp120, and inhibits syncytium formation.
This antibody is conjugated to BD Horizon BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.
Specifications
| CD4 | |
| Flow Cytometry | |
| Brilliant Violet 605 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| RPA-T4 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| L3T4; T-cell surface antigen T4/Leu-3; W3/25; CD4 antigen (p55) |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.