missing translation for 'onlineSavingsMsg'
Learn More

Anti-Human CD44 (F10-44-2), Proteintech

Mouse Monoclonal Antibody

€ 58.00 - € 179.00

Specifications

Antigen CD44
Clone F10-44-2
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30237440
View Documents
Proteintech
65063-1-IG-500UG
500 μg
€ 179.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30235301
View Documents
Proteintech
65063-1-IG-100UG
100 μg
€ 58.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CD44 antibody from Proteintech is a mouse monoclonal antibody generated with purified t cells from human lymph nodes of human CD44. This antibody recognizes human antigen. The CD44 antibody has been validated for the following applications: FC analysis.
Specifications

Specifications

CD44
0.5 mg/mL
Monoclonal
Liquid
RUO
960
Purified T cells from human lymph nodes
Primary
Store at 2-8°C. Stable for one year after shipment.
F10-44-2
Flow Cytometry
Unconjugated
Mouse
Human
CDW44; CSPG8; ECMR-III; HCELL; IN; LHR; MC56; MDU2; MDU3; MGC10468; MIC4; MUTCH-I; Pgp1
IgG2a κ
Affinity purification
Flow Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.