missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Human CD48, Brilliant Violet™ 421, Clone: TÜ145; BD
Brand: BD Biosciences 562718
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: TÜ145
Isotype: IgM, κ
Cross Reactivity Species: Hu
Applications:
Flow Cytometry
Specifications
| Human CD48 | |
| TÜ145 | |
| Mouse | |
| 100 Tests | |
| Primary | |
| IgM |
| Monoclonal | |
| Brilliant Violet 421 | |
| Affinity Purified | |
| RUO | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction