Learn More
Anti-Human CD5 (L17F12), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65200-1-IG-100UG
 View more versions of this product
Product Code. 30234440
  • € 52.00 / 100µg
Description
The CD5 antibody from Proteintech is a mouse monoclonal antibody generated with human t-cell acute lymphoblastic leukemia (t-all) cells of human CD5. This antibody recognizes human antigen. The CD5 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
CD5
Monoclonal
500 μg/mL
LEU1; T1
Human T-cell acute lymphoblastic leukemia (T-ALL) cells
100 μg
Primary
Human
Flow Antibody
IgG2a κ
Flow Cytometry
L17F12
Unconjugated
Mouse
Protein A purification
RUO
921
Store at 2-8°C. Stable for one year after shipment.