Learn More
Anti-Human CD5 (L17F12), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65200-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD5 antibody from Proteintech is a mouse monoclonal antibody generated with human t-cell acute lymphoblastic leukemia (t-all) cells of human CD5. This antibody recognizes human antigen. The CD5 antibody has been validated for the following applications: FC analysis.Specifications
| CD5 | |
| Monoclonal | |
| 500 μg/mL | |
| LEU1; T1 | |
| Human T-cell acute lymphoblastic leukemia (T-ALL) cells | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG2a κ |
| Flow Cytometry | |
| L17F12 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 921 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |