missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD5 (UCHT2), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65152-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD5 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD5. This antibody recognizes human antigen. The CD5 antibody has been validated for the following applications: FC analysis.Specifications
| CD5 | |
| Monoclonal | |
| 500 μg/mL | |
| LEU1; T1 | |
| Protein A purification | |
| RUO | |
| 921 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| UCHT2 | |
| Unconjugated | |
| Mouse | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction