missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD64 (10.1), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65253-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD64 antibody from Proteintech is a mouse monoclonal antibody generated with human rheumatoid synovial fluid cells and fibronectin-purified monocytes. of human CD64. This antibody recognizes human antigen. The CD64 antibody has been validated for the following applications: FC, ELISA analysis.Specifications
| CD64 | |
| Monoclonal | |
| 0.5 mg/mL | |
| CD64; CD64A; FCRI; FLJ18345; IGFR1 | |
| Human rheumatoid synovial fluid cells and fibronectin-purified monocytes. | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry, ELISA | |
| 10.1 | |
| Unconjugated | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 2209 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction