Learn More
Anti-Human CD73 (AD2), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65162-1-IG-500UG
 View more versions of this product
Product Code. 30237930
  • € 271.75 / 500µg
Description
The CD73 antibody from Proteintech is a mouse monoclonal antibody generated with pre-b leukemia cell line of human CD73. This antibody recognizes human antigen. The CD73 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
CD73
Monoclonal
0.5 mg/mL
CD73; E5NT; NT; NT5; NTE; eN; eNT
Pre-B leukemia cell line
500 μg
Primary
Human
Flow Antibody
IgG1 κ
Flow Cytometry
AD2
Unconjugated
Mouse
Protein A purification
RUO
4907
Store at 2-8°C. Stable for one year after shipment.