missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD85j / LILRB1 (GHI/75), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65238-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD85j / LILRB1 antibody from Proteintech is a mouse monoclonal antibody generated with human hairy cell leukemia spleen cells of human CD85j / LILRB1. This antibody recognizes human antigen. The CD85j / LILRB1 antibody has been validated for the following applications: FC analysis.Specifications
| CD85j / LILRB1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| CD85; CD85J; FLJ37515; ILT2; LIR-1; LIR1; MIR-7; MIR7 | |
| Human Hairy Cell Leukemia Spleen Cells | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG2b κ |
| Flow Cytometry | |
| GHI/75 | |
| Unconjugated | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 10859 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction