Learn More
inhibitor of DNA binding 1, dominant negative helix-loop-helix protein, Mouse, Clone: 4G11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ID1.
Brand: Abnova H00003397-M04.100ug
Description
The protein encoded by this gene is a helix-loop-helix (HLH) protein that can form heterodimers with members of the basic HLH family of transcription factors. The encoded protein has no DNA binding activity and therefore can inhibit the DNA binding and transcriptional activation ability of basic HLH proteins with which it interacts. This protein may play a role in cell growth, senescence, and differentiation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MKVASGSTATAAAGPSCALKAGKTASGAGEVVRCLSEQSVAISRCAGGAGARLPALLDEQQVNVLLYDMNGCYSRLKELVPTLPQNRKVSKVEILQHVIDYIRDLQLELNSESEVGTPGGRGLPVRAPLSTLNGEISALTAEAACVPADDRILCRSpecifications
| inhibitor of DNA binding 1, dominant negative helix-loop-helix protein | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC000613 | |
| ID1 | |
| ID1 (AAH00613.1, 1 a.a. ∼ 155 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Stem Cell Biology | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 4G11 | |
| Mouse monoclonal antibody raised against a full length recombinant ID1. | |
| ID1 | |
| ID/bHLHb24 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 3397 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.