Learn More
isocitrate dehydrogenase 2 (NADP+), mitochondrial, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant IDH2.
Brand: Abnova H00003418-A01.50uL
Description
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. Each NADP(+)-dependent isozyme is a homodimer. The protein encoded by this gene is the NADP(+)-dependent isocitrate dehydrogenase found in the mitochondria. It plays a role in intermediary metabolism and energy production. This protein may tightly associate or interact with the pyruvate dehydrogenase complex. [provided by RefSeq]
Sequence: HYREHQKGRPTSTNPIASIFAWTRGLEHRGKLDGNQDLIRFAQMLEKVCVETVESGAMTKDLAGCIHGLSNVKLNEHFLNTTDFLDTIKSNLDRALGRSpecifications
| isocitrate dehydrogenase 2 (NADP+), mitochondrial | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant IDH2. | |
| IDH2 | |
| ICD-M/IDH/IDHM/IDP/IDPM/mNADP-IDH | |
| Mouse | |
| 50 μL | |
| Yes | |
| 3418 | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002168 | |
| IDH2 | |
| IDH2 (NP_002159, 354 a.a. ∼ 451 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| This antibody cross-reacts with human IDH1. | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.