missing translation for 'onlineSavingsMsg'
Learn More

IFN-α (2b) Mouse anti-Human, NA/LE, Unlabeled, Clone: 7N4-1, BD

Product Code. 15826698
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15826698 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15826698 Supplier BD Biosciences Supplier No. 551795

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The 7N4-1 antibody reacts with human IFN-α2b and to a lesser extent with IFN-α7. It does not react with IFN-α1 nor IFN-α4. IFN-α2b is one of the three variants of IFN-α2 that have been isolated from human cell lines. IFN-α2b is the variant predominantly produced by human leukocytes. Human IFN-α2b belongs to the IFN-α class of proteins also known as leukocyte interferons. IFN-α comprises a family of related but distinct proteins with molecular weights ranging from 16-27kDa with antiviral, antiproliferative and immunomodulatory activities. The IFN-α family is composed from as many as 14 different genes. The immunogen used to generate the 7N4-1 hybridoma was E. coli-expressed recombinant human IFN-α2b. This is a neutralizing antibody.

Host Species: Mouse
Clone: 7N4-1
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: E. coli-expressed recombinant human IFN-α2b

Functional Assay, Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-α (2b)
Applications Flow Cytometry
Classification Monoclonal
Clone 7N4-1
Concentration 1.0mg/mL
Conjugate Unconjugated
Description IFNa, IFNα
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Mouse
Immunogen E. coli-expressed recombinant human IFN-α2b
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.