Learn More
IFN-Gamma Mouse anti-Human, BUV395, Clone: B27, BD
Description
The B27 monoclonal antibody specifically binds to human interferon-γ (IFN-γ). IFN-γ is a potent multifunctional cytokine that is produced by several activated cell types including NK, NKT, CD4+TCRαβ+, CD8+TCRαβ+, and TCRγδ+ T cells. IFN-γexerts its biological effects through specific binding to the high-affinity IFN-γ Receptor Complex comprised of IFN-γβ subunits. In addition to its antiviral effects, IFN-γupregulates a number of lymphoid cell functions including the antimicrobial and antitumor responses of macrophages, NK cells, and neutrophils. In addition, IFN-γcan exert strong regulatory influences on the proliferation, differentiation, and effector responses of B cell and T cell subsets. These influences can involve IFN-γ’s capacity to boost MHC class I and II expression by antigen-presenting cells as well as to direct effects on B cells and T cells themselves. Human IFN-γis a 14-18 kDa glycoprotein containing 143 amino acid residues. B27 is a neutralizing antibody. The use of B27 antibody for epitope mapping of human IFN-γ has been described. The B27 antibody has been reported not to bind to denatured IFN-γ.
Specifications
Specifications
| Antigen | IFN-Gamma |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | B27 |
| Conjugate | Brilliant Ultraviolet 395 |
| Description | IFNG; Interferon-gamma; Interferon-γ; Type II interferon; MAF |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human IFN-Gamma Recombinant Protein |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.