missing translation for 'onlineSavingsMsg'
Learn More

IFN-γ Mouse anti-Human, PE, Clone: 25723.11, BD

Product Code. 10386924
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10386924 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10386924 Supplier BD Biosciences Supplier No. 340452

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Anti-Hu–IFN-γ, clone 25723.11, is derived from the hybridization of mouse P3X-63-Ag8.653 myeloma cells with lymph node cells from BALB/c mice immunized with recombinant human IFN-γ.

Anti-Human Interferon-γ(Anti-Hu–IFN-γ) recognizes a 20- to 25-kilodalton (kd) glycoprotein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-γ
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 25723.11
Concentration 7.5μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Recombinant human IFN-γ
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.