Learn More
IFN-γ Mouse anti-Human, PE, Clone: 4S.B3, BD
Description
The 4S.B3 antibody reacts with human interferon-γ (IFN-γ). The immunogen used to generate this hybridoma was partially purified human IFN-γ obtained from supernatants of human PBMC stimulated with Staphylococcus aureus. This is a neutralizing antibody.
Host Species: Mouse
Clone: 4S.B3
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Partially purified human IFN-γ from supernatants of human PBMC stimulated with Staphylococcus aureus
Intracellular Staining
Specifications
Specifications
| Antigen | IFN-γ |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 4S.B3 |
| Concentration | 0.2mg/mL |
| Conjugate | PE |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Partially purified human IFN-γ from supernatants of human PBMC stimulated with Staphylococcus aureus |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.