missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-γ Mouse anti-Pig, PE, Clone: P2G10, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559812
This item is not returnable.
View return policy
Description
The P2G10 antibody reacts with porcine interferon-γ (IFN-γ). The immunogen used to generate the P2G10 hybridoma was recombinant pig IFN-γ.
Intracellular Staining
Specifications
| IFN-γ | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant pig IFN-γ | |
| 0.1 mg | |
| Immunology | |
| Pig | |
| IgG1 κ |
| Flow Cytometry | |
| P2G10 | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction