Learn More
anti-Ig Isotype Control Cocktail - C, Clone: MOPC-21, MOPC-21, MOPC-21, BD
Brand: BD Biosciences 558659
Additional Details : Weight : 0.01000kg
Description
Flow Cytometry
Specifications
Flow Cytometry | |
FITC/APC/PE | |
20 Tests |
MOPC-21 | |
Affinity Purified | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |