Learn More
anti-Ig Isotype Control Cocktail - C, Clone: MOPC-21, MOPC-21, MOPC-21, BD
Brand:  BD Biosciences 558659
Additional Details : Weight : 0.01000kg
Product Code. 15859558
  • € 595.00 / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
Flow Cytometry
Specifications
Specifications
Flow Cytometry
FITC/APC/PE
20 Tests
MOPC-21
Affinity Purified
Undiluted at 4°C. Protect from exposure to light. Do not freeze.