missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig κ Light Chain Mouse anti-Human, APC-H7, Clone: G20-193, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561325
This item is not returnable.
View return policy
Description
The G20-193 monoclonal antibody specifically binds to human immunoglobulin light chain, kappa (κ). It does not react with human immunoglobulin μ light chains or human immunoglobulin heavy chains.
Specifications
| Ig κ Light Chain | |
| Monoclonal | |
| APC-H7 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| G20-193 | |
| Ig, kappa Light Chain; IGKC; HCAK1; Km; Ig kappa chain C region | |
| Mouse | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction