Learn More
Ig λ Light chain Mouse anti-Human, Unlabeled, Clone: JDC-12, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555793
Description
The JDC-12 antibody reacts with human immunoglobulin light chain, lambda (λ). It does not react with κ light chain or heavy chains. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: JDC-12
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| Ig λ Light chain | |
| Monoclonal | |
| 0.5mg/mL | |
| Ig λ; IGL@; IGL; Immunoglobulin lambda locus | |
| Mouse | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| ELISA | |
| JDC-12 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.