missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig, κ Rat anti-Mouse, PE-Cy7, Clone: 187.1, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 560667
This item is not returnable.
View return policy
Description
The 187.1 monoclonal antibody specifically binds to kappa light chains of mouse immunoglobulins. The 187.1 antibody does not react with mouse μ1 or μ2 immunoglobulin lights chains or mouse immunoglobulin heavy chains.
Specifications
| Ig, κ | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2b κ secreted by MPC-11 plasmacytoma | |
| 50 μg | |
| Immunology | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| 187.1 | |
| PE-Cyanine7 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction