missing translation for 'onlineSavingsMsg'
Learn More

IgA Rat anti-Mouse, Unlabeled, Clone: C10-3, BD

Product Code. 15828808
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15828808 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15828808 Supplier BD Biosciences Supplier No. 556969

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

The C10-3 antibody reacts specifically with mouse IgA of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes.

Host Species: Rat
Clone: C10-3
Isotype: IgG1, κ
Cross Reactivity: Mouse

ELISA, Flow Cytometry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgA
Applications ELISA
Classification Monoclonal
Clone C10-3
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Pooled mouse IgA
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.