missing translation for 'onlineSavingsMsg'
Learn More

IgE Rat anti-Mouse, Brilliant Violet 786, Clone: R35-72, BD

Product Code. 15893439
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15893439 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15893439 Supplier BD Biosciences Supplier No. 564206

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

The rat anti-mouse IgE antibody (clone R35-72) reacts specifically with mouse IgE of the Igh-C [a] and Igh-C [b] haplotypes. It has been reported not to react with other Ig isotypes. Detection with the rat anti-mouse IgE antibody (clone R35-72) of surface immunoglobulin on IgE-secreting hybridoma cells has also been reported.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgE
Applications Flow Cytometry
Classification Monoclonal
Clone R35-72
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Description Igh-7; immunoglobulin heavy chain 7 (heavy chain of IgE)
Host Species Rat
Immunogen Mouse IgE (pooled)
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.