Learn More
anti-IgG1 κ, Mouse, APC-H7, Clone: MOPC-21, Isotype Control, BD
Brand: BD Biosciences 560167
Additional Details : Weight : 0.50000kg
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1, κ
Species Reactivity: Human
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
0.2mg/mL | |
Mouse | |
0.1 mg | |
Undiluted at 4°C |
MOPC-21 | |
APC-H7 | |
Affinity Purified | |
Human | |
IgG1 κ |