Learn More
anti-IgG1 κ, Mouse, Clone: 107.3, NA/LE, Isotype Control, BD
Brand: BD Biosciences 554721
Additional Details : Weight : 0.50000kg
Description
The 107.3 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human or mouse cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone 107.3 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested for endotoxin levels. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 107.3
Isotype: IgG1, κ
Immunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Functional Assay, Intracellular Staining, Isotype Control
Specifications
Functional Assay, Flow Cytometry, Western Blot | |
1mg/mL | |
Mouse | |
Affinity Purified | |
Undiluted at 4°C |
107.3 | |
Unconjugated | |
TNP-keyhole limpet hemocyanin | |
0.5 mg | |
IgG1 κ |