Learn More
anti-IgG2a, APC, Clone: X40, BD
Product Code. 15865888
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15865888 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15865888 Supplier BD Biosciences Supplier No. 340473
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Control, Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG2a
Classification Monoclonal
Clone X39
Conjugate APC
Host Species Mouse
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Isotype IgG2a