Learn More
anti-IgG2a κ, Mouse , Pacific Blue™, Clone: G155-178, BD
Brand: BD Biosciences 558118
Description
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues.
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a κImmunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
| Flow Cytometry | |
| 0.2mg/mL | |
| Mouse | |
| Affinity Purified | |
| Undiluted at 4°C |
| G155-178 | |
| Pacific Blue | |
| TNP-keyhole limpet hemocyanin | |
| 0.1 mg | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.