Learn More
anti-IgG2b κ, Rat, PE-Cy™5, Clone: A95-1, Isotype Control, BD
Brand: BD Biosciences 553990
Additional Details : Weight : 0.10000kg
Description
The A95-1 antibody has unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat, or non-human primate cells. The A95-1 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: A95-1
Isotype: IgG2b, κ
Immunogen: TNP-Keyhole Limpet Hemocyanin
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
A95-1 | |
PE-Cyanine5 | |
TNP-keyhole limpet hemocyanin | |
0.1 mg | |
IgG2b κ |