missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 Rat anti-Human and Viral, Unlabeled, Clone: JES3-9D7, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554497
This item is not returnable.
View return policy
Description
The JES3-9D7 antibody reacts with human IL-10 and viral IL-10. The immunogen used to generate the JES3-9D7 hybridoma was recombinant human IL-10 expressed in COS cells.
Specifications
| IL-10 | |
| Monoclonal | |
| 0.5mg/mL | |
| CSIF; Cytokine synthesis inhibitory factor; TGIF | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| ELISA | |
| JES3-9D7 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| COS-expressed Recombinant Human IL-10 | |
| 0.5 mg | |
| Immunology | |
| Human, Virus | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction