Learn More
IL-10 Rat anti-Mouse, Unlabeled, Clone: JES5-16E3, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 559063
Description
The JES5-16E3 antibody reacts with mouse interleukin-10 (IL-10). The immunogen used to generate the JES5-16E3 hybridoma was recombinant mouse IL-10. This is a neutralizing antibody. This antibody is routinely tested by immunocytochemistry. Other applications were tested at BD Biosciences Pharmingen during antibody development or reported in the literature.
Immunocytochemistry
Specifications
| IL-10 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant mouse IL-10 | |
| 0.25 mg | |
| Immunology | |
| Mouse | |
| IgG2b |
| Immunocytochemistry | |
| JES5-16E3 | |
| Unconjugated | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.