missing translation for 'onlineSavingsMsg'
Learn More

IL-15 Monoclonal Antibody (34559), APC, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Thermo Scientific MA523627

282.68 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.



Additional Details : Weight : 0.00010kg

Product Code. 15503322

  • € 424.00 / 100 tests
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2, recombinant mouse IL-15, or rhIL-21 is observed.

IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

IL-15
Monoclonal
APC
IL15
AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15
Mouse
Protein A/G
RUO
3600
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
34559
Buffered saline solution with BSA and 0.09% sodium azide
P40933
IL15
E. coli-derived recombinant human IL-15 Asn49-Ser162
100 Tests
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers