Learn More
IL-15 Mouse anti-Human, Biotin, Clone: G243-886, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554713
Description
The G243-886 antibody reacts with human interleukin-15 (IL-15). The immunogen used to generate the G243-886 hybridoma was recombinant human IL-15.
Host Species: Mouse
Clone: G243-886
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant Human IL-15
ELISA
Specifications
| IL-15 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human IL-15 | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 |
| ELISA | |
| G243-886 | |
| Biotin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.