Learn More
IL-15 Mouse anti-Human, Unlabeled, Clone: G243-935, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554712
Description
The G243-935 antibody reacts with human IL-15. The immunogen used to produce the G243-935 hybridoma was E. coli expressed recombinant human IL-15.
Host Species: Mouse
Clone: G243-935
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human IL-15 Recombinant Protein
ELISA
Specifications
| IL-15 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human IL-15 Recombinant Protein | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 |
| ELISA | |
| G243-935 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.