Learn More
IL-17F Mouse anti-Human, Brilliant Violet 786, Clone: O33-782, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 564265
Description
The O33-782 monoclonal antibody specifically binds to Interleukin-17F (IL-17F). IL-17F is a member of the IL-17 family of cytokines. IL-17F is encoded by the IL17F gene located in chromosome 6 (location: 6p12). IL-17F is a proinflammatory cytokine that is produced by activated T cells including differentiated CD4+ T helper 17 (Th17) cells. Activated Th17 cells can express disulfide-linked IL-17F and IL-17A homodimers as well as IL-17A/IL-17F heterodimers. These IL-17 dimers act by binding to and signaling through IL-17 receptor complexes (IL-17R). IL-17R are comprised of transmembrane IL-17RA and IL-17-RC protein subunits that are expressed by a variety of target cells including epithelial and endothelial cells, keratinocytes, fibroblasts, and granulocytes. IL-17F can induce target cells to produce proinflammatory cytokines such as IL-1 β, IL-6, G-CSF, GM-CSF, and TNF and chemokines including CXCL1/Gro-α, CXCL2/Gro-β, and CXCL8/IL-8 that attract and activate leukocytes, eg, neutrophils. Th17 and other IL-17F-producing cells play protective roles in the clearance of extracellular pathogens, including bacteria and fungi. IL-17F can also play adverse roles in inflammation associated with asthma and autoimmune diseases.
Specifications
| IL-17F | |
| Monoclonal | |
| Brilliant Violet 786 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide | |
| Human IL-17F | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry, Immunocytochemistry | |
| O33-782 | |
| Interleukin-17F; cytokine ML-1; ML-1; IL-24 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.