missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IL-2; Clone: JES6-5H4; BD
Brand: BD Biosciences 563911
This item is not returnable.
View return policy
Description
Host Species: Rat
Clone: JES6-5H4
Isotype: IgG2b
Species Reactivity: Ms
Immunogen: Mouse IL-2 Recombinant Protein
Flow Cytometry (Intracellular Staining)
Specifications
| IL-2 | |
| JES6-5H4 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse |
| Monoclonal | |
| 0.2mg/mL | |
| Mouse IL-2 Recombinant Protein | |
| 50 μg | |
| Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction