missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 Rat anti-Mouse, PE-Cy7, Clone: JES6-5H4, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 560538
This item is not returnable.
View return policy
Description
The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2) and has been reported to be neutralizing.
Host Species: Rat
Clone: JES6-5H4
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: Mouse IL-2 Recombinant Protein
Intracellular Staining
Specifications
| IL-2 | |
| Monoclonal | |
| 0.2mg/mL | |
| Il2; Interleukin-2; T-cell growth factor; TCGF | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| JES6-5H4 | |
| PE-Cyanine7 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse IL-2 Recombinant Protein | |
| 50 μg | |
| Immunology | |
| Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction