Learn More
IL-5 Rat anti-Human, NA/LE, Unlabeled, Clone: JES1-5A10, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 562175
Description
The JES1-5A10 monoclonal antibody specifically binds to the human Interleukin-5 (IL-5). IL-5 is a cytokine that is also known as T-cell replacing factor (TRF), Eosinophil differentiation factor (EDF), and B cell differentiation factor I. IL-5 is produced by T cells, mast cells, and eosinophils. IL-5 controls the generation, differentiation, activation and localization of eosinophils. IL-5 also activates and promotes the differentiation of basophils. IL-5 plays an important role in regulating cell-mediated immunity against parasitic infections and some tumors. High level IL-5 production is involved in some allergic diseases including asthma. IL-5 exerts its biological effects by binding to and signaling through high-affinity heterodimeric IL-5 receptors comprised of transmembrane IL-5 receptor alpha (IL-5R α, aka, CD125) and beta common (βc, aka, CD131) chains. The βc chain serves as the common signaling subunit for the IL-5 receptor heterodimer as well as for the heterodimeric IL-3 and GM-CSF receptors. The immunogen used to generate the JES1-5A10 hybridoma was recombinant human IL-5. The JES1-5A10 antibody has been reported to cross react with IL-5 from rhesus monkey. This is a neutralizing antibody.
Specifications
| IL-5 | |
| Monoclonal | |
| 1.0mg/mL | |
| EDF; Eosinophil differentiation factor; TRF; T-cell replacing factor | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a |
| ELISA | |
| JES1-5A10 | |
| Unconjugated | |
| No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
| Recombinant Human IL-5 | |
| 0.5 mg | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.