Learn More
IL1A, Mouse, Clone: 1F3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant IL1A.
Brand: Abnova H00003552-M02.100ug
Description
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. This cytokine is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury, and thus induces apoptosis. This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. It has been suggested that the polymorphism of these genes is associated with rheumatoid arthritis and Alzheimer's disease. [provided by RefSeq]
Sequence: KSSKDDAKITVILRISKTQLYVTAQDEDQPVLLKEMPEIPKTITGSETNLLFFWETHGTKNYFTSVAHPNLFIATKQDYWVCLAGGPPSITDFQILENQASpecifications
| IL1A | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC013142 | |
| IL1A | |
| IL1A (AAH13142, 172 a.a. ∼ 271 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1F3 | |
| Mouse monoclonal antibody raised against a partial recombinant IL1A. | |
| IL1A | |
| IL-1A/IL1/IL1-ALPHA/IL1F1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3552 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.