Learn More
IL32, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human IL32 protein.
Brand: Abnova H00009235-B01.50uL
Description
This gene encodes a member of the cytokine family. The protein contains a tyrosine sulfation site, 3 potential N-myristoylation sites, multiple putative phosphorylation sites, and an RGD cell-attachment sequence. Expression of this protein is increased after the activation of T-cells by mitogens or the activation of NK cells by IL-2. This protein induces the production of TNFalpha from macrophage cells. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: MCFPKVLSDDMKKLKARMHQAIERFYDKMQNAESGRGQVMSSLAELEDDFKEGYLETVAAYYEEQHPELTPLLEKERDGLRCRGNRSPVPDVEDPATEEPGESFCDKVMRWFQAMLQRLQTWWHGVLAWVKEKVVALVHAVQALWKQFQSFCCSLSELFMSSFQSYGAPRGDKEELTPQKCSEPQSSKSpecifications
| IL32 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human IL32 protein. | |
| IL32 | |
| IL-32alpha/IL-32beta/IL-32delta/IL-32gamma/NK4/TAIF/TAIFa/TAIFb/TAIFc/TAIFd | |
| Mouse | |
| 50 μL | |
| Yes | |
| 9235 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_001012631.1 | |
| IL32 | |
| IL32 (NP_001012649.1, 1 a.a. ∼ 188 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.