missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGB1 Monoclonal Antibody (MEM-101A), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119458
Additional Details : Weight : 0.01000kg
Description
This antibody will not cross-react with mouse.
ITGB1 Monoclonal antibody specifically detects ITGB1 in Canine, Human, Porcine samples. It is validated for Flow Cytometry.Specifications
ITGB1 | |
Monoclonal | |
APC | |
ITGB1 | |
4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CSAT antigen; ENSMUSG00000051907; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; Gm9863; gpIIa; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta 1A precursor; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin beta1D; integrin precursor; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITBG1D; ITGB1; JG22 antigen; MDF2; MSK12; RGD-receptor; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3688, 397019, 477956 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-101A | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P05556, Q9GLP0 | |
ITGB1 | |
Burkitt's lymphoma cell line Raji. | |
100 Tests | |
Primary | |
Canine, Human, Porcine | |
Antibody | |
IgG1 |