Learn More
IRAK4 Mouse anti-Human, Alexa Fluor 647, Clone: L29-525, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560315
Description
Interleukin 1 (IL-1) is a proinflammatory cytokine that mediates the host inflammatory response. IL-1 exerts its biological effects through its type I receptor (IL-1RI), which is found on most cell types. When IL-1 binds to the IL-1RI, NF-κB (transcription factor) is activated and translocates to the nucleus where it induces gene expression. IL-1-receptor-associated kinase (IRAK) has been reported to co-immunoprecipitate with IL-1RI. The IRAK family is made up of four identified Pelle homologs that regulate the activation of NF-κB and mitogen-activated protein (MAP) kinase pathways. IRAK4 is a unique member of the IRAK family because it is the closest mammalian homolog to Pelle. Also, when overexpressed, its kinase activity allows it to activate both the NF-κB and mitogen-activated protein (MAP) kinase pathways, and block the IL-1-induced activation and modification of IRAK1. IRAK4 is also able to phosphorylate IRAK1, which suggests that IRAK4 affects the early signal transduction of Toll/IL-1 receptors before the involvement of IRAK1. The L29-525 monoclonal antibody recognizes human IRAK4, regardless of phosphorylation status. The specificity of this antibody conjugate for flow cytometric analysis was validated by confirming that RNA-mediated interference (RNAi) of the specific protein reduced the staining of the cells (see figure). Furthermore, the capacity of the RNAi to down-regulate the expression of the relevant protein was confirmed by western blot analysis.
Host Species: Mouse
Clone: L29-525
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human IRAK4 Recombinant Protein
Intracellular Staining
Specifications
| IRAK4 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human IRAK4 Recombinant Protein | |
| 50 Tests | |
| Cell Biology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| L29-525 | |
| IRAK-4, REN64, Q69FE3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.