missing translation for 'onlineSavingsMsg'
Learn More

IRAK4 Mouse anti-Human, PE, Clone: L29-525, BD

Product Code. 15880089
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15880089 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15880089 Supplier BD Biosciences Supplier No. 560303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Interleukin 1 (IL-1) is a proinflammatory cytokine that mediates the host inflammatory response. IL-1 exerts its biological effects through its type I receptor (IL-1RI), which is found on most cell types. When IL-1 binds to the IL-1RI, NF-κB (transcription factor) is activated and translocates to the nucleus where it induces gene expression. IL-1-receptor-associated kinase (IRAK) has been reported to co-immunoprecipitate with IL-1RI. The IRAK family is made up of four identified Pelle homologs that regulate the activation of NF-κB and mitogen-activated protein (MAP) kinase pathways. IRAK4 is a unique member of the IRAK family because it is the closest mammalian homolog to Pelle. Also, when overexpressed, its kinase activity allows it to activate both the NF-κB and mitogen-activated protein (MAP) kinase pathways, and block the IL-1-induced activation and modification of IRAK1. IRAK4 is also able to phosphorylate IRAK1, which suggests that IRAK4 affects the early signal transduction of Toll/IL-1 receptors before the involvement of IRAK1. The L29-525 monoclonal antibody recognizes human IRAK4, regardless of phosphorylation status. The specificity of this antibody conjugate for flow cytometric analysis was validated by confirming that RNA-mediated interference (RNAi) of the specific protein reduced the staining of the cells (see figure). Furthermore, the capacity of the RNAi to down-regulate the expression of the relevant protein was confirmed by western blot analysis.

Host Species: Mouse
Clone: L29-525
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human IRAK4 Recombinant Protein

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IRAK4
Applications Flow Cytometry
Classification Monoclonal
Clone L29-525
Conjugate PE
Description IRAK-4, REN64, Q69FE3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IRAK4 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.