Learn More
ISL LIM homeobox 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human ISL1 protein.
Brand: Abnova H00003670-D01P.100ug
Description
This gene encodes a member of the LIM/homeodomain family of transcription factors. The encoded protein binds to the enhancer region of the insulin gene, among others, and may play an important role in regulating insulin gene expression. The encoded protein is central to the development of pancreatic cell lineages and may also be required for motor neuron generation. Mutations in this gene have been associated with maturity-onset diabetes of the young. [provided by RefSeq
Sequence: MGDPPKKKRLISLCVGCGNQIHDQYILRVSPDLEWHAACLKCAECNQYLDESCTCFVRDGKTYCKRDYIRLYGIKCAKCSIGFSKNDFVMRARSKVYHIECFRCVACSRQLIPGDEFALREDGLFCRADHDVVERASLGAGDPLSPLHPARPLQMAAEPISARQPALRPHVHKQPEKTTRVRTVLNEKQLHTLRTCYAANPRPDALMKEQLVEMTGLSPRVIRVWFQNKRCKDKKRSIMMKQLQQQQPNDKTNIQGMTGTPMVAASPERHDGGLQANPVEVQSYQPPWKVLSDFALQSDIDQPAFQQLVNFSEGGPGSNSTGSEVASMSSQLPDTPNSMVASPIEASpecifications
| ISL LIM homeobox 1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human ISL1 protein. | |
| ISL1 | |
| Isl-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3670 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002202 | |
| ISL1 | |
| ISL1 (NP_002193.1, 1 a.a. ∼ 346 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.