Learn More
jumonji, AT rich interactive domain 1A, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant JARID1A.
Brand: Abnova H00005927-A01.50uL
Description
The protein encoded by this gene is a ubiquitously expressed nuclear protein. It binds directly, with several other proteins, to retinoblastoma protein which regulates cell proliferation. This protein also interacts with rhombotin-2 which functions distinctly in erythropoiesis and in T-cell leukemogenesis. Rhombotin-2 is thought to either directly affect the activity of the encoded protein or may indirectly modulate the functions of the retinoblastoma protein by binding to this protein. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Sequence: KVEPEVLSTDTQTSPEPGTRMNILPKRTRRVKTQSESGDVSRNTELKKLQIFGAGPKVVGLAMGTKDKEDEVTRRRKVTNRSDAFNMQMRQRKGTLSVNFSpecifications
| jumonji, AT rich interactive domain 1A | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant JARID1A. | |
| JARID1A | |
| KDM5A/RBBP2/RBP2 | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_005056 | |
| JARID1A | |
| JARID1A (NP_005047, 191 a.a. ∼ 290 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 5927 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.