missing translation for 'onlineSavingsMsg'
Learn More

jumonji, AT rich interactive domain 2, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16160345
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16160345 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16160345 Supplier Abnova Supplier No. H00003720A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant JARID2.

This gene is an ortholog of the mouse jumonji gene, which encodes a nuclear protein essential for mouse embryogenesis, including neural tube formation. Overexpression of mouse jumonji negatively regulates cell proliferation. The jumonji proteins contain a DNA-binding domain, called an AT-rich interaction domain (ARID), and share regions of similarity with human retinoblastoma-binding protein-2 and the human SMCX protein. [provided by RefSeq

Sequence: LQLETSERRCQICQHLCYLSMVVQENENVVFCLECALRHVEKQKSCRGLKLMYRYDEEQIISLVNQICGKVSGKNGSIENCLSKPTPKRGPRKRATVDVP

Specifications

Antigen jumonji, AT rich interactive domain 2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant JARID2.
Formulation 50% glycerol
Gene JARID2
Gene Accession No. NM_004973
Gene Alias JMJ
Gene Symbols JARID2
Host Species Mouse
Immunogen JARID2 (NP_004964, 1130 a.a. ∼ 1229 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3720
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.