missing translation for 'onlineSavingsMsg'
Learn More
Learn More
jumping translocation breakpoint (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant JTB.
Brand: Abnova H00010899-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: MLAGAGRPGLPQGRHLCWLLCAFTLKLCQAEAPVQEEKLSASTSNLPCWLVEEFVVAEECSPCSNFRAKTTPECGPTGYVEKITCSSSKRNEFKSCRSALMEQRLFWKFEGAVVCVALIFACLVIIRQRQLDRKALEKVRKQIESISpecifications
| jumping translocation breakpoint | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant JTB. | |
| JTB | |
| HJTB/HSPC222/PAR/hJT | |
| Mouse | |
| 50 μL | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC000996 | |
| JTB | |
| JTB (AAH00996.1, 1 a.a. ∼ 146 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 10899 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction